missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 401130250

 View more versions of this product

Catalog No. AC401130250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium carbonate
For analysis ACS
White to Yellow
25 g
(NH4)2CO3
15, 505
Solubility in water: soluble (320g/L at 20°C)
C(=O)([O-])[O-].[NH4+].[NH4+]
96.09
96.09
ACS Reagent
58.0°C
506-87-6
Powder
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
diazanium;carbonate
517111
≥30.0% (NH3)
HDPE plastic bottle

Chemical Identifiers

506-87-6
96.09
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
517111
C(=O)([O-])[O-].[NH4+].[NH4+]
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
diazanium;carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-058-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only