Learn More
Ammonium carbonate, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196650025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium carbonate | |
p.a. | |
White | |
2.5 kg | |
(NH4)2CO3 | |
15, 505 | |
Solubility in water: soluble (320g/L at 20°C) | |
C(=O)([O-])[O-].[NH4+].[NH4+] | |
96.09 | |
96.09 | |
Analytical |
58.0°C | |
506-87-6 | |
Crystalline Powder | |
CH8N2O3 | |
MFCD00010890 | |
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate | |
PRKQVKDSMLBJBJ-UHFFFAOYSA-N | |
diazanium;carbonate | |
517111 | |
≥30.0% (NH3) | |
Plastic bottle |
Chemical Identifiers
506-87-6 | |
96.09 | |
PRKQVKDSMLBJBJ-UHFFFAOYSA-N | |
517111 | |
C(=O)([O-])[O-].[NH4+].[NH4+] |
CH8N2O3 | |
MFCD00010890 | |
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate | |
diazanium;carbonate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-058-
RUO – Research Use Only