Learn More
Ammonium cerium(IV) nitrate, 99%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423540250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Ammonium cerium(IV) nitrate | |
98.5% | |
Beige-Yellow to White | |
25 g | |
H8CeN8O18 | |
MFCD00151121 | |
15,1991 | |
Solubility in water: 1410g/L (20°C). Other solubilities: insoluble in fat, soluble in alcohol | |
548.22 | |
ACS Reagent | |
1.05 |
107.0°C to 108.0°C | |
16774-21-3 | |
Flakes or Powder | |
99% | |
(NH4)2Ce(NO3)6 | |
01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68 | |
Ceric ammonium nitrate, CAN | |
548.22 | |
99% | |
Glass bottle |
Chemical Identifiers
16774-21-3 | |
548.22 | |
Ceric ammonium nitrate, CAN |
H8CeN8O18 | |
MFCD00151121 |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 240-827-6
TSCA : TSCA
RUO – Research Use Only