Learn More
Ammonium chloride, for analysis, Thermo Scientific Chemicals
$61.26 - $494.47
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
SMILES | N.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC393182500
|
Thermo Scientific Chemicals
393182500 |
250 g | Glass bottle |
N/A
|
|
||||
AC393180010
|
Thermo Scientific Chemicals
393180010 |
1 kg | Plastic bottle |
N/A
|
|
||||
AC393180050
|
Thermo Scientific Chemicals
393180050 |
5 kg | Plastic drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
N.Cl |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 |
Specifications
340.0°C | |
White | |
250 g | |
NH3·HCl | |
15, 506 | |
Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
≥99.0% | |
Glass bottle | |
1.5200g/mL |
12125-02-9 | |
Crystalline Powder and/or Crystals | |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
53.49 | |
Analytical | |
0.01% max | |
Ammonium chloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d
GHS Signal Word: Warning
EINECSNumber : 235-186-4
RUO – Research Use Only