Learn More
Ammonium Chloride (Crystalline/Certified ACS), Fisher Chemical™
$360.08 - $2751.92
Chemical Identifiers
| CAS | 12125-02-9 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.49 |
| MDL Number | MFCD00011420 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| PubChem CID | 25517 |
| ChEBI | CHEBI:31206 |
| IUPAC Name | amine hydrochloride |
| SMILES | N.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A661-500
|
Fisher Chemical
FLA661500 |
500 g | Poly Bottle |
Each for $360.08
Case of 6 Each for $2,160.48
|
|
||||
|
A661-3
|
Thermo Fisher Scientific
FLA6613 |
3 kg | Poly Pail |
Each for $559.63
|
|
||||
|
A661-10
|
Thermo Fisher Scientific
FLA66110 |
10 kg | Poly Pail |
Each for $2,751.92
|
|
||||
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| amine hydrochloride |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 | |
| N.Cl |
Specifications
| 520°C | |
| 12125-02-9 | |
| 5.0 | |
| 500 g | |
| ClH4N | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| ≥99.5% | |
| Pass Test | |
| 1.3mbar at 160°C |
| 340°C | |
| White | |
| Solid | |
| ≥99.5% | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 25517 | |
| 53.49 | |
| Certified ACS | |
| Poly Bottle | |
| Ammonium Chloride |
Safety and Handling
WARNING!
Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:0
Instability:1