Learn More
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03912722
Description
Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium copper(II) chloride dihydrate | |
Blue | |
Crystals | |
Cl4CuH12N2O2 | |
14,514 | |
Soluble in water,acids and alcohol; Slightly soluble in ammonia. | |
[NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl | |
277.454 | |
277.46 (241.43 Anhydrous) |
110°C (decomposition) | |
10060-13-6 | |
100 g | |
MFCD00150384 | |
ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate | |
LLMDTWKBBWCWNN-UHFFFAOYSA-L | |
diazanium;tetrachlorocopper(2-);dihydrate | |
16210985 | |
1.993 g/mL |
Chemical Identifiers
10060-13-6 | |
277.454 | |
LLMDTWKBBWCWNN-UHFFFAOYSA-L | |
16210985 | |
[NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl |
Cl4CuH12N2O2 | |
MFCD00150384 | |
ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate | |
diazanium;tetrachlorocopper(2-);dihydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-690-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only