Learn More
Ammonium-d{4} deuteroxide, 99% (Isotopic), 23.0-27.0 wt% soln. in D{2}O, Thermo Scientific Chemicals
$52.04 - $52.04
Chemical Identifiers
CAS | 12168-30-8 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 40.077 |
MDL Number | MFCD00064511 |
InChI Key | VHUUQVKOLVNVRT-NSPFYZSMSA-N |
Synonym | ammonium deuteroxide, ammonium-d4 deuteroxide solution, 25 wt. % in d2o, 99 atom % d |
PubChem CID | 16211396 |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA2298003
|
Thermo Scientific Chemicals
02298003 |
1 g |
Each for $52.04
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12168-30-8 | |
40.077 | |
VHUUQVKOLVNVRT-NSPFYZSMSA-N | |
16211396 |
H5NO | |
MFCD00064511 | |
ammonium deuteroxide, ammonium-d4 deuteroxide solution, 25 wt. % in d2o, 99 atom % d | |
N.O |
Specifications
Liquid | |
1 g | |
Colorless | |
H5NO | |
MFCD00064511 | |
ammonium deuteroxide, ammonium-d4 deuteroxide solution, 25 wt. % in d2o, 99 atom % d | |
N.O | |
16211396 | |
99% | |
Ammonium-d4 deuteroxide, Isotopic |
1.334 | |
Moisture | |
12168-30-8 | |
ND4OD | |
UN2672 | |
VHUUQVKOLVNVRT-NSPFYZSMSA-N | |
40.077 | |
40.09 | |
23.0 to 27.0 wt% soln. in D2O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
EINECSNumber : 235-332-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only