missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium dichromate, ACS reagent, Thermo Scientific Chemicals
$117.62 - $344.47
Chemical Identifiers
CAS | 7789-09-5 |
---|---|
Molecular Formula | H8Cr2N2O7 |
Molecular Weight (g/mol) | 252.07 |
InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
Synonym | ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 |
PubChem CID | 24600 |
IUPAC Name | diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC446921000
|
Thermo Scientific Chemicals
446921000 |
100 g | Plastic bottle |
Each for $117.62
|
|
|||||
AC446925000
|
Thermo Scientific Chemicals
446925000 |
500 g | Plastic bottle |
Each for $344.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-09-5 | |
252.07 | |
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
H8Cr2N2O7 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
24600 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Specifications
170.0°C | |
100 g | |
(NH4)2Cr2O7 | |
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24600 | |
ACS Reagent | |
Ammonium dichromate |
7789-09-5 | |
H8Cr2N2O7 | |
15,512 | |
Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
252.07 | |
252.07 | |
Plastic bottle |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge
GHS Signal Word: Danger
EINECSNumber : 232-143-1
RUO – Research Use Only