missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium dihydrogen phosphate, 98+%, for biochemistry, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 447840010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium dihydrogen phosphate | |
7722-76-1 | |
98+% | |
NH4H2PO4 | |
15,541 | |
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone | |
[NH4+].OP(O)([O-])=O | |
115.03 | |
CHEBI:62982 | |
98+% | |
Plastic bottle |
for biochemistry | |
1kg | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03 | |
Biochemistry |
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
azanium;dihydrogen phosphate |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
[NH4+].OP(O)([O-])=O |
Safety and Handling
EINECSNumber : 231-764-5
RUO â Research Use Only