missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium dihydrogen phosphate, 98+%, for biochemistry, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 447842500

 View more versions of this product

Catalog No. AC447842500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ammonium dihydrogen phosphate
7722-76-1
98+%
NH4H2PO4
15,541
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone
[NH4+].OP(O)([O-])=O
115.03
CHEBI:62982
98+%
Plastic bottle
for biochemistry
250g
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
LFVGISIMTYGQHF-UHFFFAOYSA-N
azanium;dihydrogen phosphate
24402
115.03
Biochemistry

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
azanium;dihydrogen phosphate
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
[NH4+].OP(O)([O-])=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-764-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only