missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium dihydrogen phosphate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A152830E

 View more versions of this product

Catalog No. AAA152830E

Explore available promotions

Description

Description

Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.
Specifications

Specifications

Ammonium dihydrogen phosphate
7722-76-1
H6NO4P
14,543
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.
N.OP(O)(O)=O
115.03
CHEBI:62982
98%
1.803
198°C to 204°C
2500 g
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
LFVGISIMTYGQHF-UHFFFAOYSA-N
phosphoric acid amine
24402
115.03
Odorless

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
N.OP(O)(O)=O
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 231-764-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only