missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
$142.63 - $1990.40
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
SMILES | N.OP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC193701000
|
Thermo Scientific Chemicals
193701000 |
100 g | Plastic bottle |
Each for $142.63
|
|
||||
AC193705000
|
Thermo Scientific Chemicals
193705000 |
500 g | Plastic bottle |
Each for $507.21
|
|
||||
AC193700025
|
Thermo Scientific Chemicals
193700025 |
2.5 kg | Plastic bottle |
Each for $1,990.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
N.OP(O)(O)=O |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 |
Specifications
190.0°C | |
7722-76-1 | |
100 g | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03 | |
Trace Metal Basis | |
Ammonium dihydrogen phosphate |
White | |
Crystalline Powder | |
99.90% | |
NH4H2PO4 | |
15,541 | |
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
99.9% | |
Plastic bottle |
Safety and Handling
EINECSNumber : 231-764-5
RUO – Research Use Only