missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 98+%, ACS reagent

CAS: 12125-01-8 | FH4N | 37.04 g/mol

$61.74 - $828.78

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423290050
SDS View Documents
Thermo Scientific Chemicals
423290050
5 g Plastic bottle
Each for $61.74
Only null left
 
AC423295000
SDS View Documents
Thermo Scientific Chemicals
423295000
500 g Plastic bottle
Each for $329.37
Only null left
 
AC423290025
SDS View Documents
Thermo Scientific Chemicals
423290025
2.5 kg Plastic bottle
Each for $828.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12125-01-8
37.04
LDDQLRUQCUTJBB-UHFFFAOYSA-N
N.F
FH4N
MFCD00011423
amine hydrofluoride

Specifications

12125-01-8
Crystalline Powder
98+%
NH4F
15, 519
LDDQLRUQCUTJBB-UHFFFAOYSA-N
amine hydrofluoride
37.04
ACS Reagent
0.01% max
White
5 g
FH4N
MFCD00011423
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
N.F
37.04
≥98%
Plastic bottle
Ammonium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 235-185-9

RTECSNumber : BQ6300000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only