missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, LC-MS, Thermo Scientific™

Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument.

Supplier:  Fisher Chemical MB1221

Catalog No. MB1221



Explore available promotions

Description

Description

Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, 0.57 to 0.69g/L (Ammonium Formate, w/v), CAS Number-540-69-2, 67-56-1, 64-18-6

  • Suitable for use in LC/MS and UHPLC applications
  • Pre-mixed for ease-of-use
  • Mobile phase blend suitable for vitamin D and drugs of abuse testing  
  • Packaged in borosilicate glass bottles to prevent metal adduct formation, which can be used directly on the instrument
  • Ammonium formate- a commonly used HPLC buffer that is compatible with LC/MS
Specifications

Specifications

64.7°C
Mobile-phase solvent blend
3.5 (25°C)
1 L
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
UHPLC-MS
Borosilicate Glass Bottle
0.791 g/cm3
Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid
540-69-2,67-56-1,64-18-6
Liquid
CH4O
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
CO
32.04
10mM Ammonium Formate
128 at 20°C
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only