Learn More
Ammonium formate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401150050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium formate | |
540-69-2 | |
Crystalline Powder and/or Crystals | |
99% | |
HCO2NH4 | |
01,38; 13,230; 15,13 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
azanium;formate | |
2723923 | |
63.06 | |
Authentic |
115.0°C to 120.0°C | |
White | |
5 kg | |
CH5NO2 | |
02,8 | |
15,52 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(=O)[O-].[NH4+] | |
63.06 | |
CHEBI:63050 | |
99% | |
Plastic drum |
Chemical Identifiers
540-69-2 | |
63.06 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CHEBI:63050 | |
C(=O)[O-].[NH4+] |
CH5NO2 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
2723923 | |
azanium;formate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protect
GHS Signal Word: Warning
EINECSNumber : 208-753-9
RUO – Research Use Only