Learn More
Ammonium Formate (Crystalline/Certified ACS), Fisher Chemical
Supplier: Thermo Fisher Scientific FLA666500
Specifications
| 180°C | |
| 115°C | |
| White | |
| Solid | |
| 97 to 103 % | |
| HCOONH4 | |
| ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
| C(=O)[O-].[NH4+] | |
| 63.056 | |
| CHEBI:63050 | |
| 97 to 103% | |
| Pass Test | |
| 1.28g/mL |
| Ammonium Formate | |
| 540-69-2 | |
| 6.0 to 7.0 | |
| 500 g | |
| CH5NO2 | |
| MFCD00013103 | |
| VZTDIZULWFCMLS-UHFFFAOYSA-N | |
| azanium;formate | |
| 2723923 | |
| 63.06 | |
| Certified | |
| Poly Bottle |
Chemical Identifiers
| 540-69-2 | |
| 63.056 | |
| VZTDIZULWFCMLS-UHFFFAOYSA-N | |
| 2723923 | |
| azanium;formate |
| CH5NO2 | |
| MFCD00013103 | |
| ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
| CHEBI:63050 | |
| C(=O)[O-].[NH4+] |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:1
Instability:1