missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals

Ammonium hexafluorophosphate, 99.5%, Alfa Aesar, Quantity: 100g, CAS: 16941-11-0, Melting Point: 198 deg.C, Molecular Formula: F6H4NP, Molecular Weight: 163.003

Supplier:  Thermo Scientific Chemicals 01149514

 View more versions of this product

Catalog No. AA1149514

Explore available promotions

Description

Description

Ammonium hexafluorophosphate is used as a preservative. It is used as a starting material in the preparation of other hexafluorophosphates. Further, it is used in synthesis of organometalic compounds, especially rhodicinium hexafluorophosphate, which has inert and non-coordinating counter ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium hexafluorophosphate
16941-11-0
25 g
F6H4NP
ammonium hexafluorophosphate, ammonium phosphorus hexafluoride, ammonium hexafluorophosphate v, ammoniumhexafluorophosphate 6ci,7ci, ammoniumhexafluorophosphate 6ci,7ci, azanium hexafluorophosphate, nh4pf6, nh4pf6, ammonium hexafluoro-, ammonium hexafluoro-, e5-phosphanuide
NIZXKAYXSNUDOU-UHFFFAOYSA-O
azanium;hexafluorophosphate
9793912
Hygroscopic
198°C
Powder
99.5%
MFCD00064642
Freely soluble in water (75g/100ml). Soluble in acetone,methanol,ethanol,methyl acetate.Soluble in water.
[NH4+].F[P-](F)(F)(F)(F)F
163.003
163
2.180 g/mL

Chemical Identifiers

16941-11-0
163.003
NIZXKAYXSNUDOU-UHFFFAOYSA-O
9793912
[NH4+].F[P-](F)(F)(F)(F)F
F6H4NP
MFCD00064642
ammonium hexafluorophosphate, ammonium phosphorus hexafluoride, ammonium hexafluorophosphate v, ammoniumhexafluorophosphate 6ci,7ci, ammoniumhexafluorophosphate 6ci,7ci, azanium hexafluorophosphate, nh4pf6, nh4pf6, ammonium hexafluoro-, ammonium hexafluoro-, e5-phosphanuide
azanium;hexafluorophosphate
Safety and Handling

Safety and Handling

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 241-009-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only