Learn More
Ammonium hydrogen sulfate, 99.9% (metals basis)
CAS: 7803-63-6 | H5NO4S | 115.10 g/mol
$100.79 - $895.01
Chemical Identifiers
| CAS | 7803-63-6 |
|---|---|
| Molecular Formula | H5NO4S |
| Molecular Weight (g/mol) | 115.10 |
| MDL Number | MFCD00051073 |
| InChI Key | BIGPRXCJEDHCLP-UHFFFAOYSA-N |
| Synonym | ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4 |
| PubChem CID | 24655 |
| SMILES | [NH4+].OS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1448514
|
Thermo Scientific Chemicals
01448514 |
25 g |
Each for $100.79
|
|
|||||
|
AA1448522
|
Thermo Scientific Chemicals
01448522 |
100 g |
Each for $225.96
|
|
|||||
|
AA1448536
|
Thermo Scientific Chemicals
01448536 |
500 g |
Each for $895.01
|
|
|||||
Description
Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7803-63-6 | |
| 115.10 | |
| BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
| 24655 |
| H5NO4S | |
| MFCD00051073 | |
| ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4 | |
| [NH4+].OS([O-])(=O)=O |
Specifications
| 146.9°C | |
| Lump | |
| H5NO4S | |
| UN2506 | |
| Soluble in water (∽115.1mg/ml at 20°C). Insoluble in acetone,alcohol,and pyridine. | |
| [NH4+].OS([O-])(=O)=O | |
| 115.10 | |
| 115.1 | |
| Hygroscopic | |
| (metals basis) |
| 7803-63-6 | |
| 25 g | |
| MFCD00051073 | |
| ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4 | |
| BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
| azanium;hydrogen sulfate | |
| 24655 | |
| 99.9% | |
| Ammonium hydrogen sulfate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGEN SULFATE
EINECSNumber : 232-265-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only