missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical

Supplier:  Thermo Fisher Scientific FLA667212

Catalog No. A667-212


Explore available promotions

Description

Description

Specifications

Specifications

12
Colorless
7664-41-7,7732-18-5
H3N
MFCD00011418
2.5 L
N
17.03
CHEBI:16134
27.0 to 31.0% (NF)
Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Meets Requirements (NF)
500 hPa at 20°C
Ammonium Hydroxide
38°C
-57°C
NH4OH
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia
222
35.05
FCC/NF
Glass
0.59g/mol
Liquid

Chemical Identifiers

7664-41-7
17.03
QGZKDVFQNNGYKY-UHFFFAOYSA-N
222
ammonia
H3N
MFCD00011418
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
CHEBI:16134
N
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Accessories
Promotions

Promotions