Learn More
Ammonium Hydroxide, ACS Grade, 28.0 to 30.0%, LabChem™
Supplier: LabChem LC110501
Specifications
Ammonium Hydroxide | |
as NH3 | |
1336-21-6,7732-18-5 | |
as NH3 | |
NH4OH | |
UN2672 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
500 mL | |
N.O | |
35.05 | |
CHEBI:18219 | |
28.0 to 30.0% | |
Passes Test | |
Amber Glass | |
Liquid |
Colorless | |
27°C | |
71,29 | |
H5NO | |
MFCD00066650 | |
0.89 | |
Soluble in water | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
ACS | |
Ammonia | |
0.89 |
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
amine hydrate |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
N.O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
GHS P Statement
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
EINECSNumber : 215-647-6
Recommended Storage : Room Temperature