missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide, ACS Grade, 28.0 to 30.0%, LabChem™

Supplier:  LabChem LC110503

 View more versions of this product

Catalog No. LC110503


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide
as NH3
1336-21-6,7732-18-5
as NH3
NH4OH
UN2672
Soluble in water
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
ACS
Ammonia
0.89
Colorless
27°C
71,29
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
2.5 L
N.O
35.05
CHEBI:18219
28.0 to 30.0%
Passes Test
HDPE Bottle
Liquid

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed

Danger

EINECSNumber : 215-647-6

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions