missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide, ACS Grade, 28.0 to 30.0%, LabChem™

Supplier:  LabChem LC1105036S

 View more versions of this product

Catalog No. LC1105036S


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide
as NH3
1336-21-6,7732-18-5
as NH3
NH4OH
UN2672
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
6 x 2.5 L
N.O
35.05
CHEBI:18219
28.0 to 30.0%
Passes Test
Clear Glass
Liquid
Colorless
27°C
71,29
H5NO
MFCD00066650
0.89
Soluble in water
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
ACS
Ammonia
0.89

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 215-647-6

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions