Learn More
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
$54.60 - $180.76
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC423300250
|
Thermo Scientific Chemicals
423300250 |
25 mL |
Each for $54.60
|
|
|||||
AC423305000
|
Thermo Scientific Chemicals
423305000 |
500 mL |
Each for $84.53
|
|
|||||
AC423300025
|
Thermo Scientific Chemicals
423300025 |
2.5 L |
Each for $180.76
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
N.O |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 |
Specifications
Colorless | |
1336-21-6,7732-18-5 | |
H5NO | |
MFCD00066650 | |
0.9100g/mL | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
25 mL | |
N.O | |
35.05 | |
CHEBI:18219 | |
28.0 to 30.0% (HCl-titration (as NH3)) | |
Glass bottle | |
0.91 | |
Ammonium hydroxide |
38.0°C | |
-57.0°C | |
NH4OH | |
05,15 | |
15, 491 | |
Solubility in water: soluble. | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
ACS Reagent | |
0.002% max. | |
500hPa at 20°C |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 215-647-6
RTECSNumber : BQ9625000
TSCA : TSCA
RUO – Research Use Only