missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide, For Phenols, Certified, 0.50N ±0.01N (0.5M), LabChem™
Supplier: LabChem LC110701
Specifications
Ammonium Hydroxide | |
For Phenols | |
99.19,0.81 | |
NH4OH | |
0.50N ±0.01N (0.5M) | |
Soluble in water | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
Passes Test | |
Poly Bottle | |
Liquid |
Colorless | |
1336-21-6,7732-18-5 | |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
500 mL | |
N.O | |
35.05 | |
CHEBI:18219 | |
Certified | |
Ammonia | |
Traceable to NIST |
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
amine hydrate |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
N.O |
Safety and Handling
GHS H Statement
Harmful to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage : Room Temperature