missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide (Optima™), Fisher Chemical™

Supplier:  Seastar Chemicals Inc S020701TFIJ05

 View more versions of this product

Catalog No. A4701


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide
1336-21-6
NH4OH
0.920g/cm³
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
HDPE Bottle
Liquid
Colorless
H5NO
MFCD00066650
15, 491
1 L
N.O
35.05
CHEBI:18219
OPTIMA™
501 hPa at 50°C

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions