missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, Puriss., 30-33% NH3 in H2O, Honeywell Fluka™
Puriss., 30-33% NH3 in H2O
$174.70 - $922.60
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| SMILES | N.O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 |
Specifications
| 1336-21-6 | |
| NH4OH | |
| 30 to 33% NH3 in H2O | |
| 3587154 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| azanium;hydroxide | |
| 35.05 | |
| CHEBI:18219 | |
| puriss. | |
| Ammonium Hydroxide Solution |
| H5NO | |
| MFCD00066650 | |
| 2672 | |
| 0.9g/mL at 25°C | |
| 1 L | |
| N.O | |
| 0.27 | |
| 14923 | |
| 35.05g/mol | |
| Glass Bottle |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314-H335-H410
DOTInformation : 8 / PGIII