missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide Solution, puriss. p.a. plus, Honeywell Fluka™
Puriss. p.a. plus,≥25% NH3 in H2O
$279.55 - $279.55
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000297
|
Honeywell-Fluka
170931L |
1 L |
N/A
|
|
|||||
Description
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
N.O |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 |
Specifications
1336-21-6 | |
NH4OH | |
2672 | |
0.91g/mL at 20°C, 0.9g/mL at 25°C | |
1 L | |
N.O | |
0.27 | |
14923 | |
35.05g/mol | |
puriss. p.a. plus | |
Ammonium Hydroxide Solution |
H5NO | |
MFCD00066650 | |
3587154 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
35.05 | |
CHEBI:18219 | |
≥25% (NH3 in H2O) | |
Plastic Bottle |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
H302-H314-H410