missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Iodide, (ACS Reagent Grade), MP Biomedicals

Supplier:  MP Biomedicals Inc 0215249680

 View more versions of this product

Catalog No. ICN15249680


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Iodide
100 g
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide
25487
144.9
12027-06-4
H4IN
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
[NH4+].[I-]
144.94
CHEBI:63040
ACS Reagent

Chemical Identifiers

12027-06-4
144.94
UKFWSNCTAHXBQN-UHFFFAOYSA-N
25487
ammonium iodide
H4IN
MFCD00011424
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
CHEBI:63040
[NH4+].[I-]
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions