Learn More
Ammonium iron(III) oxalate trihydrate, 98%, Thermo Scientific Chemicals
$82.47 - $269.60
Chemical Identifiers
CAS | 13268-42-3 |
---|---|
Molecular Formula | C6H18FeN3O15 |
Molecular Weight (g/mol) | 428.06 |
MDL Number | MFCD00150137 |
InChI Key | ARJSTCBPHIPXIJ-UHFFFAOYSA-K |
Synonym | Ferric ammonium oxalate; Iron(III) ammonium oxalate |
IUPAC Name | triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4âµ.4âµ]tridecane-5,5,5-tris(ylium) trihydrate |
SMILES | [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2141122
|
Thermo Scientific Chemicals
B2141122 |
100 g |
Each for $82.47
|
|
|||||
AAB2141136
|
Thermo Scientific Chemicals
B2141136 |
500 g |
Each for $269.60
|
|
|||||
Description
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
13268-42-3 | |
428.06 | |
ARJSTCBPHIPXIJ-UHFFFAOYSA-K | |
triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4âµ.4âµ]tridecane-5,5,5-tris(ylium) trihydrate |
C6H18FeN3O15 | |
MFCD00150137 | |
Ferric ammonium oxalate; Iron(III) ammonium oxalate | |
[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4 |
Specifications
13268-42-3 | |
C6H18FeN3O15 | |
UN3288 | |
Ferric ammonium oxalate; Iron(III) ammonium oxalate | |
ARJSTCBPHIPXIJ-UHFFFAOYSA-K | |
triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4âµ.4âµ]tridecane-5,5,5-tris(ylium) trihydrate | |
428.07 (374.02 Anhydrous) | |
Hygroscopic; Light sensitive | |
Ammonium iron(III) oxalate trihydrate |
100 g | |
MFCD00150137 | |
14,517 | |
Soluble in water. Insoluble in ethanol. | |
[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4 | |
428.06 | |
98% | |
1.78 |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 238-090-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only