missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iron(III) sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

$99.15 - $341.49

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1610536
View Documents
Thermo Scientific Chemicals
A1610536
500 g
Each for $99.15
 
AAA161050E
View Documents
Thermo Scientific Chemicals
A161050E
2500 g
Each for $341.49
 
Description

Description

Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

Solubility
Soluble in water (1240 mg/ml at 25°C) and 1% (v/v) aqueous HCl (100 mg/ml). Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

7783-83-7
482.176
LCPUDZUWZDSKMX-UHFFFAOYSA-K
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
FeH28NO20S2
MFCD00150004
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
azanium;iron(3+);disulfate;dodecahydrate

Specifications

39°C to 41°C
500 g
NH4Fe(SO4)2·12H2O
14,521
Soluble in water (1240mg/ml at 25°C) and 1% (v/v) aqueous HCl (100mg/ml). Insoluble in alcohol.
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
482.18 (266.00 Anhydrous)
Odorless
Ammonium iron(III) sulfate dodecahydrate
7783-83-7
FeH28NO20S2
MFCD00150004
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
≥98%
1.71
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 233-382-4

RTECSNumber : WS5900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only