missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Oxalate Monohydrate, greaterthan equal99%, ACS, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219140890

 View more versions of this product

Catalog No. ICN19140890

Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Oxalate Monohydrate
White
Powder
≥99%
Number MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
diazanium;oxalate;hydrate
516808
ACS
Monohydrate
6009-70-7
500 g
C2H10N2O5
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
142.111
142.1

Chemical Identifiers

6009-70-7
142.111
MSMNVXKYCPHLLN-UHFFFAOYSA-N
516808
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
C2H10N2O5
Number MFCD00149694
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
diazanium;oxalate;hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-135-4

SDS
Documents

Documents

Promotions

Promotions