missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate, Monohydrate, Purified, Spectrum™ Chemical

$516.29 - $7208.29

Chemical Identifiers

CAS 6009-70-7
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608732
View Documents
Spectrum Chemical Mfg Cor
A1221500GM
500 g Poly Bottle
Each for $516.29
Only null left
 
18608733
View Documents
Spectrum Chemical Mfg Cor
A122125KG
2.5 kg Poly Bottle
Each for $1,436.85
Only null left
 
18608734
View Documents
Spectrum Chemical Mfg Cor
A122112KG
12 kg Poly Pail
Each for $4,144.15
Only null left
 
18608735
View Documents
Spectrum Chemical Mfg Cor
A122150KGBL
50 kg Fiber Drum
Each for $7,208.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6009-70-7

Specifications

6009-70-7
500 g
Poly Bottle
1
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.