missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Persulfate, Molecular Biology Reagent, MP Biomedicals™

Ammonium Persulfate, Molecular Biology Reagent is a polymerization catalyst used with TEMED for polyacrylamide gel formation.

$58.45 - $364.35

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19398825
View Documents
MP Biomedicals Inc
0219398825
25 g
Each for $58.45
 
ICN19398890
View Documents
MP Biomedicals Inc
0219398890
500 g
Each for $364.35
 
Description

Description

  • Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
  • APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry.
  • Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, and the decolorizing and deodorizing of oils.
Specifications

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

120°C (Decomposition)
7727-54-0
25 g
MFCD00003390
Soluble in water, 83.5 g/100 g water at 25°C
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
228.19
228.18
Molecular Biology
Ammonium persulfate
White
Crystalline Powder
H8N2O8S2
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
ROOXNKNUYICQNP-UHFFFAOYSA-N
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
62648
≥98%
1.98 g/cm3 at 68°F; Bulk Density: 951 to 1050 kg/m3
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.