Learn More
Ammonium phosphate, dibasic, 99+%, for analysis
CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
$110.48 - $746.04
Identifiants chimiques
| CAS | 7783-28-0 |
|---|---|
| Molecular Formula | H9N2O4P |
| Molecular Weight (g/mol) | 132.06 |
| MDL Number | MFCD00010891 |
| InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
| Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
| PubChem CID | 24540 |
| ChEBI | CHEBI:63051 |
| IUPAC Name | phosphoric acid diamine |
| SMILES | N.N.OP(O)(O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC201822500
|
Thermo Scientific Chemicals
201822500 |
250 g | Plastic bottle |
chaque for $110.48
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC201825000
|
Thermo Scientific Chemicals
201825000 |
500 g | Plastic bottle |
chaque for $186.33
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC201820025
|
Thermo Scientific Chemicals
201820025 |
2.5 kg | Plastic bottle |
chaque for $746.04
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Identifiants chimiques
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| phosphoric acid diamine |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 | |
| N.N.OP(O)(O)=O |
Spécifications
| 7783-28-0 | |
| 7.9 to 8.3 (5% soln. at 20°C) | |
| 250 g | |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| phosphoric acid diamine | |
| 24540 | |
| 132.06 | |
| Analytical | |
| 1.61 |
| White | |
| Adhering Crystals | |
| 99+% | |
| (NH4)2HPO4 | |
| 15, 540 | |
| Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| ≥99% | |
| Plastic bottle | |
| Ammonium phosphate, dibasic, p.a. |
Sécurité et manipulation
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 231-987-8
missing translation for 'tsca' : TSCA
RUO – Research Use Only