missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Phosphate, Monobasic, Crystal, ACS, 98%, Spectrum™ Chemical
$159.15 - $2546.59
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
IUPAC Name | phosphoric acid amine |
SMILES | N.OP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18608765
|
Spectrum Chemical Mfg Cor
A123525GM |
25 g | Poly Bottle |
Each for $159.15
|
|
||||
18608766
|
Spectrum Chemical Mfg Cor
A1235125GM |
125 g | Poly Bottle |
Each for $317.26
|
|
||||
18608767
|
Spectrum Chemical Mfg Cor
A1235500GM |
500 g | Poly Bottle |
Each for $505.85
|
|
||||
18608768
|
Spectrum Chemical Mfg Cor
A123525KG |
2.5 kg | Poly Bottle |
Each for $1,388.53
|
|
||||
18608769
|
Spectrum Chemical Mfg Cor
A123512KG |
12 kg | Poly Pail |
Each for $2,546.59
|
|
||||
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
N.OP(O)(O)=O |
H6NO4P | |
MFCD00003396 | |
phosphoric acid amine |
Specifications
7722-76-1 | |
3.8 to 4.4 | |
H6NO4P | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
98% | |
Poly Bottle |
1 | |
25 g | |
MFCD00003396 | |
N.OP(O)(O)=O | |
115.03 | |
ACS |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.