missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium succinate, Thermo Scientific™

Chemical Identifiers

CAS 2226-88-2
Molecular Formula C4H12N2O4
Molecular Weight (g/mol) 152.15
InChI Key NHJPVZLSLOHJDM-UHFFFAOYSA-N
IUPAC Name diammonium butanedioate
SMILES [NH4+].[NH4+].[O-]C(=O)CCC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466860050
View Documents
Thermo Scientific Chemicals
466860050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466860250
View Documents
Thermo Scientific Chemicals
466860250
25 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2226-88-2
152.15
diammonium butanedioate
C4H12N2O4
NHJPVZLSLOHJDM-UHFFFAOYSA-N
[NH4+].[NH4+].[O-]C(=O)CCC([O-])=O

Specifications

2226-88-2
C4H12N2O4
[NH4+].[NH4+].[O-]C(=O)CCC([O-])=O
152.15
Ammonium succinate
5 g
NHJPVZLSLOHJDM-UHFFFAOYSA-N
diammonium butanedioate
152.12
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only