missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate, ACS, USP, Pyrogen Free, Macco Organiques

Macco products offered by Spectrum Chemical are suitable for biopharmaceutical production.

Supplier:  Spectrum Chemical Mfg Cor 50704000120

Catalog No. 18614839


Explore available promotions

Description

Description

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP
Specifications

Specifications

Colorless or White
4.5 to 6
12 kg
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
99 to 100.5%
Plastic Pail
7783-20-2
Crystals or Granules
H8N2O4S
One gram is soluble in about 1.5 mL of water. It is soluble in alcohol
N.N.OS(O)(=O)=O
132.13
ACS/USP
0.005%

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
N.N.OS(O)(=O)=O
H8N2O4S
MFCD00003391
sulfuric acid diamine
Videos
SDS
Documents

Documents

Promotions

Promotions