missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

A1245, 7783-20-2, (NH4)2SO4

Supplier:  Spectrum Chemical Mfg Cor A1245125GM

 View more versions of this product

Catalog No. 18600656

Explore available promotions

Description

Description

Spectrum™ Chemical Ammonium Sulfate, Crystal, Reagent, ACS is an inorganic salt that is very soluble in water Most often used as a soil fertilizer to release minute amounts of acid. It is also used in biochemistry as a precipitant to purify protein, used as a food additive and flame retardant. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

7783-20-2
5.0-6.0
H8N2O4S
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
99%
Poly Bottle
100%
125 g
MFCD00003391
N.N.OS(O)(=O)=O
132.13
ACS
0.005%

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
N.N.OS(O)(=O)=O
H8N2O4S
MFCD00003391
sulfuric acid diamine
SDS
Documents

Documents

Promotions

Promotions