missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
A1245, 7783-20-2, (NH4)2SO4
Supplier: Spectrum Chemical Mfg Cor A124550KGBLPL
Description
Spectrum™ Chemical Ammonium Sulfate, Crystal, Reagent, ACS is an inorganic salt that is very soluble in water Most often used as a soil fertilizer to release minute amounts of acid. It is also used in biochemistry as a precipitant to purify protein, used as a food additive and flame retardant. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Specifications
7783-20-2 | |
5.0-6.0 | |
H8N2O4S | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
99% | |
Poly Drum |
100% | |
50 kg | |
MFCD00003391 | |
N.N.OS(O)(=O)=O | |
132.13 | |
ACS | |
0.005% |
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
sulfuric acid diamine |