missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, For Radium, Certified, 10.0%(w/v) ±0.5%(w/v), LabChem™

Supplier:  LabChem LC113001

Catalog No. LC113001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Sulfate
For Radium
90.51,9.49
(NH4)2SO4
1.05g/mL
Soluble in water
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
10.0% ±0.5% (w/v)
Passes Test
1.05g/mL
1.1cSt
500 mL
Colorless
7783-20-2,7732-18-5
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
6097028
132.13
Certified
Poly Bottle
Traceable to NIST
Gaseous Ammonia; Sulfur compounds
Liquid

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
sulfuric acid diamine
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
N.N.OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions