missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate (Granular/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA7023

 View more versions of this product

Catalog No. A702-3



Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Sulfate
7783-20-2
5 to 6
3 kg
H8N2O4S
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
6097028
132.14
Certified ACS
Poly Bottle
280°C
Colorless
Solid
≥99.0 %
(NH4)2SO4
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
≥99.0%
Pass Test

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
sulfuric acid diamine
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
N.N.OS(O)(=O)=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions