missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Granular, FCC, 99-100.5%, Spectrum™ Chemical
AM185, 7783-20-2, (NH4)2SO4
$250.70 - $4,092.57
Chemical Identifiers
| CAS | 7783-20-2 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.13 |
| MDL Number | MFCD00003391 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| IUPAC Name | sulfuric acid diamine |
| SMILES | N.N.OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600665
|
Spectrum Chemical Mfg Cor
AM185500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18600668
|
Spectrum Chemical Mfg Cor
AM185100LBBL |
100 lb. | Fiber Drum |
N/A
|
|
||||
|
18600666
|
Spectrum Chemical Mfg Cor
AM18525KG |
2.5 kg | Poly Bottle |
N/A
|
|
||||
|
18600667
|
Spectrum Chemical Mfg Cor
AM18512KG |
12 kg | Poly Pail |
N/A
|
|
||||