missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Sulfide, Light Solution, Reagent, 20-24%, Spectrum™ Chemical
A1250, 12135-76-1, (NH4)2S
Supplier: Spectrum Chemical Mfg Cor A125025LTPV
Description
Spectrum™ Chemical Ammonium Sulfide, Light Solution, Reagent , is an unstable salt with the formula (NH4)2S. It has a very strong odor, and the Reagent grade means it is used in the laboratory as an analytical testing agent. Common uses of AMMONIUM SULFIDE, LIGHT SOLUTION, REAGENT are industrial applications including photo developing, textile manufacturing and others.Specifications
12135-76-1,7732-18-5 | |
2.5 L | |
MFCD00010892 | |
UXXGWUQNRMPNKH-UHFFFAOYSA-N | |
diamine sulfane | |
20 to 24% | |
Poly-Coated Glass Bottle |
80% | |
H8N2S | |
UN2683 | |
N.N.S | |
68.14 | |
Reagent | |
0.04% |
Chemical Identifiers
12135-76-1 | |
68.14 | |
UXXGWUQNRMPNKH-UHFFFAOYSA-N | |
N.N.S |
H8N2S | |
MFCD00010892 | |
diamine sulfane |