missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium tartrate, 98.8%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219504290

 View more versions of this product

Catalog No. ICN19504290


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium tartrate
3164-29-2
500 g
C4H12N2O6
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
184.15
CHEBI:63075
1.601g/mL
White
Crystalline Powder
0.988
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
13652278
184.1

Chemical Identifiers

3164-29-2
184.15
NGPGDYLVALNKEG-OLXYHTOASA-N
13652278
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
C4H12N2O6
MFCD00013073
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
CHEBI:63075
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Videos
SDS Documents

Documents

Promotions

Promotions