Learn More
Ammonium tetrafluoroborate, 99.999% (metals basis), Thermo Scientific Chemicals
$64.85 - $531.56
Chemical Identifiers
CAS | 13826-83-0 |
---|---|
Molecular Formula | BF4H4N |
Molecular Weight (g/mol) | 104.84 |
MDL Number | MFCD06245785 |
InChI Key | PDTKOBRZPAIMRD-UHFFFAOYSA-O |
Synonym | ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p |
PubChem CID | 9964072 |
IUPAC Name | ammonium tetrafluoroboranuide |
SMILES | [NH4+].F[B-](F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4405506
|
Thermo Scientific Chemicals
04405506 |
5 g |
Each for $64.85
|
|
|||||
AA4405514
|
Thermo Scientific Chemicals
04405514 |
25 g |
Each for $174.38
|
|
|||||
AA4405522
|
Thermo Scientific Chemicals
04405522 |
100 g |
Each for $531.56
|
|
|||||
Description
Ammonium fluoroborate is used as an analytical reagent. It is also applied in the textile printing and dyeing industry and used as a catalyst for resin finishing. It is mainly used for the high temperature fluxing action required by the metals industry; catalyst; in flame retardants and acts as a solid lubricant in cutting-oil emulsions for aluminum rolling and forming.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13826-83-0 | |
104.84 | |
PDTKOBRZPAIMRD-UHFFFAOYSA-O | |
9964072 | |
[NH4+].F[B-](F)(F)F |
BF4H4N | |
MFCD06245785 | |
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p | |
ammonium tetrafluoroboranuide |
Specifications
220°C (sublimation) | |
White | |
5 g | |
MFCD06245785 | |
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p | |
PDTKOBRZPAIMRD-UHFFFAOYSA-O | |
ammonium tetrafluoroboranuide | |
9964072 | |
99.999% | |
Hygroscopic | |
Ammonium tetrafluoroborate |
13826-83-0 | |
Crystalline | |
BF4H4N | |
UN3260 | |
Soluble in water at 16°C 25g/L. | |
[NH4+].F[B-](F)(F)F | |
104.84 | |
104.84 | |
Ammonia-like | |
1.871 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
H314-H318-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 237-531-4
RTECSNumber : BQ6100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only