missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical FLA709500

 View more versions of this product

Catalog No. A709-500



Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Thiocyanate
1762-95-4
5 to 6
500 g
CH4N2S
MFCD00011428
SOIFLUNRINLCBN-UHFFFAOYSA-N
azanium;thiocyanate
15666
≥97.5%
Pass Test
1.300g/cm³
149°C
White
Solid
≥97.5 %
NH4SCN
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
C(#N)[S-].[NH4+]
76.117
76.12
Certified ACS
Poly Bottle

Chemical Identifiers

1762-95-4
76.117
SOIFLUNRINLCBN-UHFFFAOYSA-N
15666
C(#N)[S-].[NH4+]
CH4N2S
MFCD00011428
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
azanium;thiocyanate
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Contact with acids liberates toxic gas. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:1
Instability:1

SDS Documents

Documents

Promotions

Promotions