missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium thioglycolate, 70 wt.% solution in water, Thermo Scientific Chemicals

Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5

$62.92 - $141.39

Specifications

Chemical Name or Material Ammonium thioglycolate
Color Colorless
Name Note 70 wt.% solution in water
CAS 5421-46-5,7732-18-5
Boiling Point 115.0°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384682500
View Documents
Thermo Scientific Chemicals
384682500
250 mL Glass bottle
Each for $62.92
 
AC384680010
View Documents
Thermo Scientific Chemicals
384680010
1 L Glass bottle
Each for $141.39
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium thioglycolate
70 wt.% solution in water
115.0°C
C2H7NO2S
1.2200g/mL
Solubility in water: completely soluble.
C(C(=O)[O-])S.[NH4+]
109.143
109.15
ammonium: positive, mercaptane: positive
1.22
Solution
Colorless
5421-46-5,7732-18-5
-20.0°C
MFCD00137451
ammonium thioglycolate, ammonium mercaptoacetate, acetic acid, mercapto-, monoammonium salt, thiofaco a-50, ammonium thioglycollate, thioglycolic acid ammonium salt, usaf mo-2, unii-6p9e788vzz, thioglycollic acid, ammonium salt, ammonium 2-mercaptoacetate
ZZTCCAPMZLDHFM-UHFFFAOYSA-N
azanium;2-sulfanylacetate
21534
68.0 to 72.0%
Glass bottle
5.8 to 6.4
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only