missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium thiosulfate, 96%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 391665000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Ammonium thiosulfate | |
Extra Pure | |
7783-18-8 | |
500 g | |
H8N2O3S2 | |
MFCD00015965 | |
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate | |
148.21 | |
96% | |
Plastic bottle |
150.0°C | |
White | |
Crystalline Powder | |
95% min. (Iodimetry) | |
(NH4)2S2O3 | |
15, 559 | |
Solubility in water: 63.1% (20°C) - 71.0% (70°C). Other solubilities: insoluble in alcohol and ether | |
148.21 | |
Pure |
Chemical Identifiers
7783-18-8 | |
148.21 | |
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate |
H8N2O3S2 | |
MFCD00015965 |
Safety and Handling
EINECSNumber : 231-982-
RUO – Research Use Only