missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium titanyl oxalate monohydrate, 98%, pure, Thermo Scientific Chemicals

$161.78 - $161.78

Chemical Identifiers

CAS 10580-03-7
Molecular Formula C4H8N2O9Ti
Molecular Weight (g/mol) 275.98
MDL Number MFCD00149620
InChI Key VBUNWMBOVJSSMH-UHFFFAOYSA-L
Synonym Ammonium bis(oxalato)oxotitanate(IV)
IUPAC Name diammonium oxotitaniumbis(ylium) dioxalate
SMILES [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC212730100
SDS View Documents
Thermo Scientific Chemicals
212730100
10 g Glass Bottle
Each for $161.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10580-03-7
275.98
VBUNWMBOVJSSMH-UHFFFAOYSA-L
diammonium oxotitaniumbis(ylium) dioxalate
C4H8N2O9Ti
MFCD00149620
Ammonium bis(oxalato)oxotitanate(IV)
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

White to Beige
Crystalline Powder and/or Crystals
98%
(NH4)2TiO(C2O4)2·H2O
10,584
Solubility in water: soluble
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
275.98
98%
Authentic
Ammonium titanyl oxalate monohydrate
10580-03-7
10 g
C4H8N2O9Ti
MFCD00149620
Ammonium bis(oxalato)oxotitanate(IV)
VBUNWMBOVJSSMH-UHFFFAOYSA-L
diammonium oxotitaniumbis(ylium) dioxalate
294.02
Pure
Glass Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only