AMP, 0.2M buffer soln., pH 9.5, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals J63988AE
 View more versions of this product
Catalog No. AAJ63988AE
Explore available promotions
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
124-68-5
0.2 M
Liquid
C4H11NO
MFCD00008051
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methylpropan-1-ol
Colorless
9.5
100 mL
Odorless
2-Amino-2-methyl-1-propanol
CC(C)(N)CO
89.14